missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSK1/RPS6KA5, p Ser376 Antibody (2F3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26393-100ul
This item is not returnable.
View return policy
Description
MSK1/RPS6KA5 Monoclonal antibody specifically detects MSK1/RPS6KA5 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| MSK1/RPS6KA5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 2F3 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
| A synthesized peptide derived from Human MSK1/RPS6KA5 [p Ser376] [UniProt O75582] | |
| 100 μL | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 9252 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction