missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MS4A14 Recombinant Rabbit Monoclonal Antibody (24GB3545)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | MS4A14 |
|---|---|
| Clone | 24GB3545 |
| Concentration | 0.25 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276255
![]() |
Invitrogen™
MA557083 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276463
|
Invitrogen™
MA557084 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MS4A14 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
May be involved in signal transduction as a component of a multimeric receptor complex.Specifications
| MS4A14 | |
| 0.25 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96JA4 | |
| 383435, 84689 | |
| A synthesized peptide derived from human MS4A14 (aa 150-280). | |
| Antibody |
| 24GB3545 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| MS4A14 | |
| membrane spanning 4-domains A14; membrane-spanning 4-domains subfamily A member 14; membrane-spanning 4-domains, subfamily A, member 14; membrane-spanning 4-domains, subfamily A, member 16; MS4A13; MS4A13 protein; MS4A14; MS4A16; NYD-SP21; testes development-related NYD-SP21; testis development protein NYD-SP21 | |
| MS4A14 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title