missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MS4A14 Recombinant Rabbit Monoclonal Antibody (24GB3545)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557084

 View more versions of this product

Product Code. 30276463

  • £126.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MS4A14 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

May be involved in signal transduction as a component of a multimeric receptor complex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MS4A14
Recombinant Monoclonal
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96JA4
MS4A14
A synthesized peptide derived from human MS4A14 (aa 150-280).
20 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot, Immunocytochemistry
24GB3545
Unconjugated
MS4A14
membrane spanning 4-domains A14; membrane-spanning 4-domains subfamily A member 14; membrane-spanning 4-domains, subfamily A, member 14; membrane-spanning 4-domains, subfamily A, member 16; MS4A13; MS4A13 protein; MS4A14; MS4A16; NYD-SP21; testes development-related NYD-SP21; testis development protein NYD-SP21
Rabbit
Affinity chromatography
RUO
383435, 84689
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.