missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A14 Antibody (S02-0B8), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | MS4A14 |
|---|---|
| Clone | S02-0B8 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654943
|
Novus Biologicals
NBP3-19700-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639513
|
Novus Biologicals
NBP3-19700-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MS4A14 Monoclonal antibody specifically detects MS4A14 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MS4A14 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 84689 | |
| IgG | |
| Affinity purified |
| S02-0B8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DKFZp434H092, FLJ32856, membrane-spanning 4-domains subfamily A member 14, membrane-spanning 4-domains, subfamily A, member 14, membrane-spanning 4-domains, subfamily A, member 16, MGC104289, MGC49828, MS4A13, MS4A13 protein, MS4A16, NYD-SP21, testes development-related NYD-SP21, Testis development protein NYD-SP21 | |
| Recombinant protein of human MS4A14 (Uniprot # Q96JA4) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title