missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A14 Antibody (S02-0B8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19700-100ul
This item is not returnable.
View return policy
Description
MS4A14 Monoclonal antibody specifically detects MS4A14 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MS4A14 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84689 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S02-0B8 | |
| Unconjugated | |
| DKFZp434H092, FLJ32856, membrane-spanning 4-domains subfamily A member 14, membrane-spanning 4-domains, subfamily A, member 14, membrane-spanning 4-domains, subfamily A, member 16, MGC104289, MGC49828, MS4A13, MS4A13 protein, MS4A16, NYD-SP21, testes development-related NYD-SP21, Testis development protein NYD-SP21 | |
| Recombinant protein of human MS4A14 (Uniprot # Q96JA4) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction