missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15225-1-AP-150UL
This item is not returnable.
View return policy
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S12P family. The encoded protein is a key component of the ribosomal small subunit and controls the decoding fidelity and susceptibility to aminoglycoside antibiotics. The gene for mitochondrial seryl-tRNA synthetase is located upstream and adjacent to this gene, and both genes are possible candidates for the autosomal domit deafness gene (DFNA4). Splice variants that differ in the 5′ UTR have been found for this gene; all three variants encode the same protein.Specifications
| MRPS12 | |
| Polyclonal | |
| Unconjugated | |
| MRPS12 | |
| MPR S12, MRP S12, MRPS12, MT RPS12, RPMS12, RPS12, RPSM12, S12mt | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24030, 292758, 6183 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15235, O35680 | |
| MRPS12 | |
| MRPS12 Fusion Protein Ag7442 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction