missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54647
This item is not returnable.
View return policy
Description
MRPS12 Polyclonal specifically detects MRPS12 in Human samples. It is validated for Western Blot.
Specifications
| MRPS12 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 28S ribosomal protein S12, mitochondrial, mitochondrial ribosomal protein S12, MPR-S12, MRP-S12, MT-RPS12, ribosomal protein, mitochondrial, S12, RPMS12, RPS12, RPSM12S12mt | |
| Rabbit | |
| 12 kDa | |
| 100 μL | |
| Primary | |
| Bovine: 86%; Guinea pig: 86%; Equine: 79%. | |
| Human, Rat, Bovine, Canine, Equine, Guinea Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O15235 | |
| MRPS12 | |
| Synthetic peptides corresponding to MRPS12(mitochondrial ribosomal protein S12) The peptide sequence was selected from the N terminal of MRPS12. Peptide sequence LVPRLWATCSMATLNQMHRLGPPKRPPRKLGPTEGRPQLKGVVLCTFTRK. | |
| Affinity purified | |
| RUO | |
| 6183 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction