missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRO Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79844
This item is not returnable.
View return policy
Description
MRO Polyclonal specifically detects MRO in Human samples. It is validated for Western Blot.
Specifications
| MRO | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| B29FLJ30140, beside the Ma29 deletion, C18orf3, chromosome 18 open reading frame 3, maestro, Male-specific transcription in the developing reproductive organs, Protein B29, protein maestro | |
| Rabbit | |
| 30 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Dog: 85%; Horse: 85%; Bovine: 85%; Rabbit: 85%; Rat: 77%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_001120648 | |
| MRO | |
| Synthetic peptide directed towards the C terminal of human MROThe immunogen for this antibody is MRO. Peptide sequence VAKACKTTFQACSPYLKLKEEYSFQSEEDQRNTKLYQQLSHYHPEILQFF. | |
| Affinity purified | |
| RUO | |
| 83876 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction