missing translation for 'onlineSavingsMsg'
Learn More

Mre11 Rabbit anti-Human, Mouse, Clone: 8Q2B3, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £386.00

Specifications

Antigen Mre11
Clone 8Q2B3
Dilution Western Blot 1:500 - 1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18300015
View Documents
Novus Biologicals
NBP3-16359-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18316595
View Documents
Novus Biologicals
NBP3-16359-100UL
100 μg
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mre11 Monoclonal antibody specifically detects Mre11 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

Mre11
Western Blot 1:500 - 1:2000
Monoclonal
Purified
RUO
Human, Mouse
ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human Mre11 (P49959).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
8Q2B3
Western Blot
Unconjugated
Rabbit
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase
PBS, 0.05% BSA, 50% glycerol, pH7.3
4361
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato