missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mre11 Rabbit anti-Human, Mouse, Clone: 8Q2B3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16359-20UL
This item is not returnable.
View return policy
Description
Mre11 Monoclonal antibody specifically detects Mre11 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Mre11 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 8Q2B3 | |
| Western Blot 1:500 - 1:2000 | |
| ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human Mre11 (P49959). | |
| 20 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase | |
| 4361 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction