missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £443.00
Specifications
| Antigen | MRAP |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18472161
|
Novus Biologicals
NBP1-88941-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264758
|
Novus Biologicals
NBP1-88941 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRAP Polyclonal specifically detects MRAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MRAP | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 56246 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MANGTNASAPYYSYEYYLDYLDLIPVDEKKLKAHKHS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| B27GCCD2, C21orf61chromosome 21 open reading frame 61, FALPFGD2, Fat cell-specific low molecular weight protein, Fat tissue-specific low MW protein, melanocortin 2 receptor accessory protein, melanocortin-2 receptor accessory protein | |
| MRAP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title