missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88941-25ul
This item is not returnable.
View return policy
Beskrivning
MRAP Polyclonal specifically detects MRAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| MRAP | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| B27GCCD2, C21orf61chromosome 21 open reading frame 61, FALPFGD2, Fat cell-specific low molecular weight protein, Fat tissue-specific low MW protein, melanocortin 2 receptor accessory protein, melanocortin-2 receptor accessory protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56246 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MRAP | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MANGTNASAPYYSYEYYLDYLDLIPVDEKKLKAHKHS | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering