missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPPED2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MPPED2 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16881394
|
Proteintech
13270-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16871394
|
Proteintech
13270-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Displays low metallophosphoesterase activity (in vitro). May play a role in the development of the nervous system.Specifications
| MPPED2 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| B1WBP0, Q15777, Q9CZJ0 | |
| 362185, 744, 77015 | |
| MPPED2 Fusion Protein Ag3963 | |
| Primary | |
| -20°C | |
| MPPED2 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 239FB, C11orf8, FAM1B, Fetal brain protein 239, Metallophosphoesterase MPPED2, MPPED2 | |
| Mpped2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title