missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPPED2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13270-1-AP-150UL
This item is not returnable.
View return policy
Description
Displays low metallophosphoesterase activity (in vitro). May play a role in the development of the nervous system.Specifications
| MPPED2 | |
| Polyclonal | |
| Unconjugated | |
| MPPED2 | |
| 239FB, C11orf8, FAM1B, Fetal brain protein 239, Metallophosphoesterase MPPED2, MPPED2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362185, 744, 77015 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B1WBP0, Q15777, Q9CZJ0 | |
| Mpped2 | |
| MPPED2 Fusion Protein Ag3963 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction