missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MPP5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

£133.00 - £397.00

Specifications

Antigen MPP5
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17720684
GREENER_CHOICE
View Documents
Invitrogen™
PA5119276
200 μL
£397.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17743164
View Documents
Invitrogen™
PA5121766
20 μL
£133.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MPP5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

MPP5 (Membrane Protein, Palmitoylated 5) plays a critical role in neurogenesis and hematopoiesis. This protein is crucial for establishing and maintaining cell polarity in epithelial cells and neurons. It is involved in the Par-3 complex, which contributes to maintaining tight junctions in epithelial cells. Mutations in MPP5 have been associated with severe neurologic impairments, including global developmental delay (GDD) and behavioral changes. In animal models, MPP5 is highly involved in the early differentiation of multipotent progenitors and supports long-term lineage-specific contributions, primarily in myeloid and lymphoid cells. MPP5 variants disrupt normal cellular function, leading to profound developmental and functional abnormalities.
Specifications

Specifications

MPP5
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Mouse
MPP5
3830420B02Rik; AI255216; AI644496; FLJ12615; MAGUK p55 subfamily member 5; membrane palmitoylated protein 5; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 5); membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5); MPP5; palmitoylated 5; PALS1; protein associated with Lin Seven 1; protein associated with Lin-7 1; stardust
MPP5
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q8N3R9, Q9JLB2
56217, 64398
Recombinant protein Membrane Protein, Palmitoylated 5. The antigen corresponds to amino acid range 455-675 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.