missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPHOSPH1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | MPHOSPH1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665540
|
Novus Biologicals
NBP2-94707-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672461
|
Novus Biologicals
NBP2-94707-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MPHOSPH1 Polyclonal antibody specifically detects MPHOSPH1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MPHOSPH1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 9585 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cancer/testis antigen 90, CT90MPP-1, DKFZp434B0435, EC 6.3.5.5, kinesin family member 20B, Kinesin-related motor interacting with PIN1, KRMP1kinesin-like protein KIF20B, M-phase phosphoprotein 1mitotic kinesin-like protein, MPHOSPH1EC 2.1.1, MPP1DKFZp434P0810 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1500 of human KIF20B (NP_057279.2). KHLLQLKEEEEETNRQETEKLKEELSASSARTQNLKADLQRKEEDYADLKEKLTDAKKQIKQVQKEVSVMRDEDKLLRIKINELEKKKNQCSQELDMKQRTIQQLKEQLNNQKVEEAIQQYERACKDLNVKEKIIEDMRMTLEEQEQTQVEQDQVLEAKLEEVERLATELEKWKEKCNDLETKNNQRSNKEHENNTDVLGKLTNLQDELQESEQKYNADRKKWLEEKMMLITQAKEAENIRNKEMKKYAEDRERFFKQQNEMEILTAQLTEKDSDLQKWREERDQLVAALEIQLKALISSN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title