missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPHOSPH1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94707-0.02ml
This item is not returnable.
View return policy
Description
MPHOSPH1 Polyclonal antibody specifically detects MPHOSPH1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MPHOSPH1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| Cancer/testis antigen 90, CT90MPP-1, DKFZp434B0435, EC 6.3.5.5, kinesin family member 20B, Kinesin-related motor interacting with PIN1, KRMP1kinesin-like protein KIF20B, M-phase phosphoprotein 1mitotic kinesin-like protein, MPHOSPH1EC 2.1.1, MPP1DKFZp434P0810 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1500 of human KIF20B (NP_057279.2). KHLLQLKEEEEETNRQETEKLKEELSASSARTQNLKADLQRKEEDYADLKEKLTDAKKQIKQVQKEVSVMRDEDKLLRIKINELEKKKNQCSQELDMKQRTIQQLKEQLNNQKVEEAIQQYERACKDLNVKEKIIEDMRMTLEEQEQTQVEQDQVLEAKLEEVERLATELEKWKEKCNDLETKNNQRSNKEHENNTDVLGKLTNLQDELQESEQKYNADRKKWLEEKMMLITQAKEAENIRNKEMKKYAEDRERFFKQQNEMEILTAQLTEKDSDLQKWREERDQLVAALEIQLKALISSN | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 9585 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction