missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protease, from S. aureus strain V8

Product Code. 11434211 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 11434211

Brand: MP Biomedicals™ 0215197210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for selective cleavage of proteins for amino acid sequence determination or peptide mapping

  • Unit Definition: One unit will hydrolyze casein to change A280 0.001 per minute, pH 7.8, at 37°C.
  • Extinction Coefficient: (E1%) E1%280 = 4.26
  • Solubility: Soluble in aqueous buffers (such as 10 mM Tris, pH 7.8 at 1 mg/mL).
  • CAS Number: 66676-43-5
  • Molecular Weight: 27,700 (sedimentation equilibrium)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Used for selective cleavage of Proteins for Amino Acid sequence determination or Peptide mapping
pH Range 4 and 7.8
Sample Type Staphylococcus aureus V8 Strain
Content And Storage -20°C
Form Powder
Quantity 10 mg
Product Type Protease
compliance-icons compliance-icons
Product Identifier
  • Protease
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Protease
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.