missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Neuraminidase

Brand:  MP Biomedicals™ 0215384605

Code : NEW

Additional Details : Weight : 1.36080kg

 View more versions of this product

Product Code. 11496145

  • £1321.82 / 5 units
Estimated Shipment: 28-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Activity 60 units/mg protein for NAN-lactose >25 units/mg protein for bovine submaxillary mucin >20 units/mg protein for colominic acid Protein is determined by the method of Lowry et al. with bovine albumin as a standard

Specifications

Specifications

≥60 U/mg
5
4°C
5 U
Lyophilized Powder
White
Used to cleave N-Acetyl Neuraminic Acid from a variety of Glycoproteins
Arthrobacter ureafaciens
Neuraminidase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers