missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protease (S. Aureus) V.8

Codice prodotto. 11493616 Sfoglia Tutto MP Biomedicals Prodotti
Cambia vista
Click to view available options
Quantity:
5 mg
Dimensione della confezione:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
11493616 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11493616 Fornitore MP Biomedicals™ N. del fornitore 08399001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Protease (S. Aureus) V.8, also called endoproteinase Glu-C, Glu-C protease, staphylococcal serine proteinase. Widely used for protein sequencing, this enzyme specifically cleaves peptide bonds on the COOH terminal side of either aspartic or glutamic acids.

  • Protease V.8 specifically cleaves peptide bonds on the COOH terminal side of either aspartic or glutamic acids
  • In the presence of ammonium, the specificity is limited to glutamic sites
  • Used in a study to investigate immobilized residue-specific endoproteinases for protein sequencing
  • Protease V8 was also used in a study to assess enzymatic cleaning of inorganic ultrafiltration membranes used for whey protein fractionation
  • Provides:
    • Robust activity, greater than 915 units/mg protein
    • Specific cleavage for glutamic acid in ammonium buffer
    • Selective digestion at both glutamic and aspartic acid residues in phosphate buffer at pH 7.8
    • Cleavage activity remains in the presence of many denaturing agent, such as SDS, urea and guanidine-HCl
    • Lyophilized format for longer stability and easier storage
TRUSTED_SUSTAINABILITY

Specifica

For Use With (Application) Used in a study to investigate immobilized residue-specific endoproteinases for Protein sequencing
pH Range 4 and 7.8
Sample Type Staphylococcus aureus
Content And Storage 2°C to 8°C
Form Lyophilized
Quantity 5 mg
Product Type Protease
compliance-icons compliance-icons
Avvertenza
  • Danger
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.