missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOX1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£150.00
Specifications
| Antigen | MOX1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10-20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
MOX1 Polyclonal specifically detects MOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| MOX1 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_004518 | |
| 4222 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human MOX1. The immunogen is located within amino acids 140 - 190 of MOX1. Amino Acid Squence: SRRRKESSDNQENRGKPEG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| homeobox protein MOX-1, mesenchyme homeobox 1MOX1mesenchyme homeo box 1 | |
| MEOX1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title