missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOX1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76302-0.025mg
This item is not returnable.
View return policy
Description
MOX1 Polyclonal specifically detects MOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| MOX1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| homeobox protein MOX-1, mesenchyme homeobox 1MOX1mesenchyme homeo box 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4222 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
| NP_004518 | |
| MEOX1 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human MOX1. The immunogen is located within amino acids 140 - 190 of MOX1. Amino Acid Squence: SRRRKESSDNQENRGKPEG | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction