missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse WARP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£156.00 - £413.00
Specifications
| Antigen | WARP/VWA1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778207
![]() |
R&D Systems
AF4927-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18009741
|
R&D Systems
AF4927 |
100 μg |
£413.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WARP/VWA1 Polyclonal specifically detects WARP/VWA1 in Mouse samples. It is validated for Western Blot.Specifications
| WARP/VWA1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| DKFZp761O051, von Willebrand factor A domain containing 1, von Willebrand factor A domain-containing protein 1, von Willebrand factor A domain-related protein, VWA1, WARP | |
| VWA1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8R2Z5 | |
| 64856 | |
| Mouse myeloma cell line NS0-derived recombinant mouse WARP isoform 1 Arg19-Pro415 Accession # Q8R2Z5 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse WARP in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human WARP is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title