missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse WARP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4927-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
WARP/VWA1 Polyclonal specifically detects WARP/VWA1 in Mouse samples. It is validated for Western Blot.
Specifica
| WARP/VWA1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp761O051, von Willebrand factor A domain containing 1, von Willebrand factor A domain-containing protein 1, von Willebrand factor A domain-related protein, VWA1, WARP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64856 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q8R2Z5 | |
| VWA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse WARP isoform 1 Arg19-Pro415 Accession # Q8R2Z5 | |
| 25 μg | |
| Primary | |
| Detects mouse WARP in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human WARP is observed. | |
| Mouse | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto