missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VE-Statin Isoform 1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £352.00
Specifications
| Antigen | VE-Statin/EGFL7 |
|---|---|
| Clone | 362907 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18775098
![]() |
R&D Systems
MAB3089-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18153011
![]() |
R&D Systems
MAB3089 |
100 μg |
£352.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-Statin/EGFL7 Monoclonal specifically detects VE-Statin/EGFL7 in Mouse samples. It is validated for Western Blot.Specifications
| VE-Statin/EGFL7 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9QXT5 | |
| 51162 | |
| E. coli-derived recombinant mouse VE-Statin Isoform 1 Thr21-Leu275 Accession # Q9QXT5 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse VE-Statin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human VE-Statin is observed. |
| 362907 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1 | |
| EGFL7 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title