missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VE-Statin Isoform 1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3089-SP
This item is not returnable.
View return policy
Description
VE-Statin/EGFL7 Monoclonal specifically detects VE-Statin/EGFL7 in Mouse samples. It is validated for Western Blot.
Specifications
| VE-Statin/EGFL7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9QXT5 | |
| EGFL7 | |
| E. coli-derived recombinant mouse VE-Statin Isoform 1 Thr21-Leu275 Accession # Q9QXT5 | |
| 25 μg | |
| Primary | |
| Detects mouse VE-Statin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human VE-Statin is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 362907 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51162 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction