missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Urotensin-II R Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
£218.00 - £382.00
Specifications
| Antigen | Urotensin-II R/GPR14 |
|---|---|
| Clone | 443707 |
| Dilution | Flow Cytometry 0.2 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18742108
|
R&D Systems
FAB9245R-025UG |
25 μg |
£218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753857
![]() |
R&D Systems
FAB9245R-100UG |
100 μg |
£382.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Urotensin-II R/GPR14 Monoclonal specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry.Specifications
| Urotensin-II R/GPR14 | |
| Flow Cytometry 0.2 ug/10^6 cells | |
| Monoclonal | |
| Rat | |
| Mouse | |
| G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R | |
| Mouse Urotensin-II R synthetic peptide | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 443707 | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| 2837 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse Urotensin-II R. Stains mouse Urotensin-II R transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title