missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Urotensin-II R Alexa Fluor 647-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB9245R-025UG
This item is not returnable.
View return policy
Description
Urotensin-II R/GPR14 Monoclonal specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Urotensin-II R/GPR14 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2837 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 443707 | |
| Flow Cytometry 0.2 ug/10^6 cells | |
| G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R | |
| Mouse Urotensin-II R synthetic peptide | |
| 25 μg | |
| Primary | |
| Detects mouse Urotensin-II R. Stains mouse Urotensin-II R transfectants but not irrelevant transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction