missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Tsukushi/TSK Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £346.00
Specifications
| Antigen | Tsukushi/TSK |
|---|---|
| Clone | 462310 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18769867
![]() |
R&D Systems
MAB6376-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18129970
|
R&D Systems
MAB6376 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tsukushi/TSK Monoclonal specifically detects Tsukushi/TSK in Mouse samples. It is validated for Western Blot.Specifications
| Tsukushi/TSK | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8CBR6 | |
| 25987 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tsukushi/TSK Thr18-Leu354 Accession # Q8CBR6 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects mouse Tsukushi/TSK in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human Tsukushi/TSK is observed. |
| 462310 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E2IG4, E2IG4E2-induced gene 4 protein, leucine rich repeat containing 54, Leucine-rich repeat-containing protein 54, LRRC54, TSKtsukushi homolog, TSKU, Tsukushi, tsukushi small leucine rich proteoglycan homolog (Xenopus laevis), tsukushi, small leucine rich proteoglycan, tsukushin | |
| TSKU | |
| IgG2c | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title