missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Tsukushi/TSK Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB6376-SP
This item is not returnable.
View return policy
Description
Tsukushi/TSK Monoclonal specifically detects Tsukushi/TSK in Mouse samples. It is validated for Western Blot.
Specifications
| Tsukushi/TSK | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8CBR6 | |
| TSKU | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tsukushi/TSK Thr18-Leu354 Accession # Q8CBR6 | |
| 25 μg | |
| Primary | |
| Detects mouse Tsukushi/TSK in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human Tsukushi/TSK is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 462310 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E2IG4, E2IG4E2-induced gene 4 protein, leucine rich repeat containing 54, Leucine-rich repeat-containing protein 54, LRRC54, TSKtsukushi homolog, TSKU, Tsukushi, tsukushi small leucine rich proteoglycan homolog (Xenopus laevis), tsukushi, small leucine rich proteoglycan, tsukushin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25987 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction