Learn More
Invitrogen™ TSLP Mouse Uncoated ELISA Kit with Plates
Engineered for the accurate and precise measurement of mouse TSLP in serum, and tissue culture supernatant samples.
£173.00 - £392.00
Specifications
Accession Number | Q9JIE6 |
---|---|
Assay Range | 16-2,000 pg/mL |
Assay Sensitivity | 16 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15551157
|
Invitrogen™
88-7490-22 |
2 x 96 Tests |
£173.00
Pack of 2 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
15561157
|
Invitrogen™
88-7490-86 |
10 x 96 Tests |
£392.00
Pack of 10 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
Description
The Mouse TSLP Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse TSLP. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Mouse TSLP ELISA Ready-SET-Go! reagent set contains all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assaysSpecifications
Q9JIE6 | |
16 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
IL-2,TCGF,lymphokine | |
HRP | |
Serum, 100 μL; Supernatant, 100 μL | |
Thymic Stromal-Derived Lymphopoietin | |
1 hr. 45 min. |
16-2,000 pg/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
53603 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Coating Buffer: 10X PBS ELISA Coating Buffer, Assay Diluent: 5X Concentrated, Detection Enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Solution, Certificate of Analysis: Lot-specific instructions for the dilution of antibodies and standards, 96-Well Plates: Corning Costar flat-bottom plates (included with Cat. Nos. ending in -22 and -86) | |
RUO | |
2°C to 8°C | |
Mouse | |
24 hr. 30 min. |
For Research Use Only