Learn More
Invitrogen™ Mouse TSLP Uncoated ELISA Kit with Plates
Description
The Mouse TSLP Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse TSLP. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Accession Number | Q9JIE6 |
| Assay Range | 16-2000 pg/mL |
| Assay Sensitivity | 16 pg/mL |
| Content And Storage | Capture Antibody: Pre-titrated, purified antibody Detection Antibody: Pre-titrated, biotin-conjugated antibody Standard: Recombinant cytokine for generating standard curve and calibrating samples Coating Buffer: 10X PBS ELISA Coating Buffer Assay Diluent: 5X Concentrated Detection Enzyme: Pre-titrated Avidin-HRP Substrate Solution: Tetramethylbenzidine (TMB) Solution Certificate of Analysis: Lot-specific instructions for the dilution of antibodies and standards 96-Well Plates: Corning Costar flat-bottom plates (included with Cat. Nos. ending in -22 and -86) |
| Conjugate | HRP |
| Immunoassay Kit Format | Uncoated ELISA Kit with Plates |
| Product Type | ELISA |
| Sample Type | Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.