missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TREML1/TLT-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£171.00 - £487.00
Specifications
| Antigen | TREML1/TLT-1 |
|---|---|
| Clone | 268515 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726249
![]() |
R&D Systems
MAB24241-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18193251
![]() |
R&D Systems
MAB24241 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TREML1/TLT-1 Monoclonal specifically detects TREML1/TLT-1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| TREML1/TLT-1 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dJ238O23.3, GLTL1825, PRO3438, TLT1, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
| TREML1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 268515 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| NP_082039 | |
| 340205 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREML1/TLT-1 Gly18-Cys178 Accession # NP_082039 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TREML1/TLT-1 in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TREML1 or rhTREML2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title