missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TREML1/TLT-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB24241-SP
This item is not returnable.
View return policy
Description
TREML1/TLT-1 Monoclonal specifically detects TREML1/TLT-1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| TREML1/TLT-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dJ238O23.3, GLTL1825, PRO3438, TLT1, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 340205 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 268515 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_082039 | |
| TREML1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREML1/TLT-1 Gly18-Cys178 Accession # NP_082039 | |
| 25 μg | |
| Primary | |
| Detects mouse TREML1/TLT-1 in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TREML1 or rhTREML2 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction