missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TORC1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £353.00
Specifications
| Antigen | TORC1 |
|---|---|
| Clone | 505531 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783129
![]() |
R&D Systems
MAB5978-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149610
|
R&D Systems
MAB5978 |
100 μg |
£353.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TORC1 Monoclonal specifically detects TORC1 in Mouse samples. It is validated for Immunohistochemistry.Specifications
| TORC1 | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q68ED7 | |
| 23373 | |
| E. coli-derived recombinant mouse TORC1 Gln384-Gln471 Accession # Q68ED7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TORC1 in direct ELISAs. |
| 505531 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CREB regulated transcription coactivator 1, CRTC1, FLJ14027WAMTP1, KIAA0616Transducer of regulated cAMP response element-binding protein 1, MECT1, MECT1CREB-regulated transcription coactivator 1, mucoepidermoid carcinoma translocated 1, Mucoepidermoid carcinoma translocated protein 1, TORC1TORC-1, Transducer of CREB protein 1, transducer of regulated cAMP response element-binding protein (CREB) 1, WAMTP1 | |
| CRTC1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title