missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TORC1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB5978
This item is not returnable.
View return policy
Description
TORC1 Monoclonal specifically detects TORC1 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| TORC1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q68ED7 | |
| CRTC1 | |
| E. coli-derived recombinant mouse TORC1 Gln384-Gln471 Accession # Q68ED7 | |
| 100 μg | |
| Primary | |
| Detects mouse TORC1 in direct ELISAs. | |
| Mouse | |
| Purified |
| Immunohistochemistry | |
| 505531 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CREB regulated transcription coactivator 1, CRTC1, FLJ14027WAMTP1, KIAA0616Transducer of regulated cAMP response element-binding protein 1, MECT1, MECT1CREB-regulated transcription coactivator 1, mucoepidermoid carcinoma translocated 1, Mucoepidermoid carcinoma translocated protein 1, TORC1TORC-1, Transducer of CREB protein 1, transducer of regulated cAMP response element-binding protein (CREB) 1, WAMTP1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23373 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction