missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TGF-beta 2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£171.00 - £1735.00
Specifications
| Antigen | TGF-beta 2 |
|---|---|
| Clone | 771213 |
| Dilution | Neutralization 0.15-0.75 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792919
![]() |
R&D Systems
MAB7346-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706488
![]() |
R&D Systems
MAB7346-100 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18737119
![]() |
R&D Systems
MAB7346-500 |
500 μg |
£1735.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Mouse samples. It is validated for Neutralization.Specifications
| TGF-beta 2 | |
| Neutralization 0.15-0.75 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P27090 | |
| 7042 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TGF-beta 2 Ala303-Ser414 Accession # P27090 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects mouse TGF-beta 2 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) TGF-beta 2 and rhTGF-beta 3 is observed. |
| 771213 | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
| TGFB2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title