missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TGF-beta 2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB7346-SP
This item is not returnable.
View return policy
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Mouse samples. It is validated for Neutralization.
Specifications
| TGF-beta 2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGFB2, TGFbeta 2, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7042 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization | |
| 771213 | |
| Neutralization 0.15-0.75 ug/mL | |
| P27090 | |
| TGFB2 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TGF-beta 2 Ala303-Ser414 Accession # P27090 | |
| 25 μg | |
| Primary | |
| Detects mouse TGF-beta 2 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) TGF-beta 2 and rhTGF-beta 3 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction