missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-F Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£167.00 - £1795.00
Specifications
| Antigen | Siglec-F |
|---|---|
| Clone | 238047 |
| Concentration | LYOPH |
| Dilution | Neutralization 0.1-0.5 ug/mL |
| Applications | Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18727188
![]() |
R&D Systems
MAB17061-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754137
![]() |
R&D Systems
MAB17061-100 |
100 μg |
£447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741797
![]() |
R&D Systems
MAB17061-500 |
500 μg |
£1795.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-F Monoclonal specifically detects Siglec-F in Mouse samples. It is validated for Neutralization.Specifications
| Siglec-F | |
| LYOPH | |
| Neutralization | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q920G3 | |
| 27181 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 Accession # Q920G3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Siglec-F in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Siglec-2 or -3, or recombinant human Siglec-5, -7, -9, -10, or -11 is observed. |
| 238047 | |
| Neutralization 0.1-0.5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF | |
| Siglecf | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title