missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-F Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB17061-500
This item is not returnable.
View return policy
Description
Siglec-F Monoclonal specifically detects Siglec-F in Mouse samples. It is validated for Neutralization.
Specifications
| Siglec-F | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.1-0.5 ug/mL | |
| Q920G3 | |
| Siglecf | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 Accession # Q920G3 | |
| 500 μg | |
| Primary | |
| Detects mouse Siglec-F in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Siglec-2 or -3, or recombinant human Siglec-5, -7, -9, -10, or -11 is observed. | |
| Mouse | |
| Purified |
| Neutralization | |
| 238047 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27181 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction