missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SIGIRR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1092-SP
This item is not returnable.
View return policy
Description
SIGIRR Polyclonal specifically detects SIGIRR in Mouse samples. It is validated for Western Blot.
Specifications
| SIGIRR | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1R8, single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8, TIR8MGC110992, Toll/interleukin-1 receptor 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 59307 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9JLZ8 | |
| SIGIRR | |
| Mouse myeloma cell line NS0-derived recombinant mouse SIGIRR Met1-His117 Accession # Q9JLZ8 | |
| 25 μg | |
| Primary | |
| Detects mouse SIGIRR in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) SIGIRR, rhIL-1 R9, recombinant mouse (rm) IL-18 R beta , rmIL-18 R, and recombinant rat IL-1 R6 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction