missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SIGIRR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
£168.00 - £451.00
Specifications
| Antigen | SIGIRR |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737397
![]() |
R&D Systems
AF1092-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18022742
|
R&D Systems
AF1092 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIGIRR Polyclonal specifically detects SIGIRR in Mouse samples. It is validated for Western Blot.Specifications
| SIGIRR | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| IL-1R8, single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8, TIR8MGC110992, Toll/interleukin-1 receptor 8 | |
| SIGIRR | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9JLZ8 | |
| 59307 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SIGIRR Met1-His117 Accession # Q9JLZ8 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse SIGIRR in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) SIGIRR, rhIL-1 R9, recombinant mouse (rm) IL-18 R beta , rmIL-18 R, and recombinant rat IL-1 R6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title