missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Serpin E2/PN1 Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £440.00
Specifications
| Antigen | Serpin E2/PN1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.1 ug/mL |
| Applications | Western Blot, Immunoprecipitation, Neutralization |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18796357
![]() |
R&D Systems
AF2175-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18082812
![]() |
R&D Systems
AF2175 |
100 μg |
£440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin E2/PN1 Polyclonal specifically detects Serpin E2/PN1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
| Serpin E2/PN1 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
| SERPINE2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| RUO | |
| Q07235 | |
| 5270 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Serpin E2/PN1 Ser20-Pro397 Accession # Q07235 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Serpin E2/PN1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin I2, recombinant mouse (rm) Serpin C1, rmSerpin D1, and rmSerpin F2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title