missing translation for 'onlineSavingsMsg'
Learn More

Mouse Serpin E2/PN1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18082812
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18082812 100 μg 100µg
18796357 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18082812 Supplier R&D Systems Supplier No. AF2175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Serpin E2/PN1 Polyclonal specifically detects Serpin E2/PN1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin E2/PN1
Applications Western Blot, Immunoprecipitation, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.1 ug/mL
Gene Accession No. Q07235
Gene Alias GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2
Gene Symbols SERPINE2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Serpin E2/PN1 Ser20-Pro397 Accession # Q07235
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5270
Test Specificity Detects mouse Serpin E2/PN1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin I2, recombinant mouse (rm) Serpin C1, rmSerpin D1, and rmSerpin F2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.