missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Sca-1/Ly6 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
£280.00 - £489.00
Specifications
| Antigen | Sca-1/Ly6 |
|---|---|
| Clone | 177228 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721709
![]() |
R&D Systems
FAB1226A-025 |
25 Tests |
£280.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766047
![]() |
R&D Systems
FAB1226A-100 |
100 Tests |
£489.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sca-1/Ly6 Monoclonal specifically detects Sca-1/Ly6 in Mouse samples. It is validated for Flow Cytometry.Specifications
| Sca-1/Ly6 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| C6orf23, G6D, Ly6, Ly-6, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25, Sca1 | |
| Ly6a | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse Sca-1/Ly6 in direct ELISAs. |
| 177228 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| CAA28351 | |
| 58530 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele Leu27-Gly119 Accession # CAA28351 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title