missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Sca-1/Ly6 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1226A-100
This item is not returnable.
View return policy
Description
Sca-1/Ly6 Monoclonal specifically detects Sca-1/Ly6 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Sca-1/Ly6 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| C6orf23, G6D, Ly6, Ly-6, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25, Sca1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58530 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 177228 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CAA28351 | |
| Ly6a | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele Leu27-Gly119 Accession # CAA28351 | |
| 100 Tests | |
| Primary | |
| Detects mouse Sca-1/Ly6 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction