missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat P-Selectin/CD62P Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
£187.00 - £519.00
Specifications
| Antigen | P-Selectin/CD62P |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18712898
![]() |
R&D Systems
AF737-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18028541
|
R&D Systems
AF737 |
100 μg |
£519.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P-Selectin/CD62P Polyclonal specifically detects P-Selectin/CD62P in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| P-Selectin/CD62P | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse, Rat | |
| CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP | |
| SELP | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q01102 | |
| 6403 | |
| Mouse myeloma cell line NS0-derived recombinant mouse P-Selectin/CD62P Trp42-Ala709 Accession # Q01102 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse P-Selectin/CD62P in direct ELISAs. Detects mouse and rat P-Selectin/CD62P in Western blot. In direct ELISAs, less than 15% cross-reactivity with recombinant human P-Selectin/CD62P is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title