missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat P-Selectin/CD62P Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF737
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Polyclonal specifically detects P-Selectin/CD62P in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| P-Selectin/CD62P | |
| Polyclonal | |
| Unconjugated | |
| Q01102 | |
| SELP | |
| Mouse myeloma cell line NS0-derived recombinant mouse P-Selectin/CD62P Trp42-Ala709 Accession # Q01102 | |
| 100 μg | |
| Primary | |
| Detects mouse P-Selectin/CD62P in direct ELISAs. Detects mouse and rat P-Selectin/CD62P in Western blot. In direct ELISAs, less than 15% cross-reactivity with recombinant human P-Selectin/CD62P is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
| CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6403 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction