missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat ICAM-5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
£181.00 - £460.00
Specifications
| Antigen | ICAM-5 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755909
![]() |
R&D Systems
AF1173-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18023372
![]() |
R&D Systems
AF1173 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICAM-5 Polyclonal specifically detects ICAM-5 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| ICAM-5 | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse, Rat | |
| ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
| ICAM5 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Q60625 | |
| 7087 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5 Leu31-Arg828 (Pro47Arg) Accession # Q60625 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse ICAM-5 in direct ELISAs. Detects mouse and rat ICAM-5 in Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ICAM-5 and recombinant mouse (rm) ICAM-2 is observed, 5% cross-reactivity with recombinant rat ICAM-1, rhICAM-1, and rhICAM-3 is observed and less than 1% cross-reactivity with rmICAM-1 and rmVCAM-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title