missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat ICAM-5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Marca: R&D Systems AF1173
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ICAM-5 Polyclonal specifically detects ICAM-5 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifica
| ICAM-5 | |
| Polyclonal | |
| Unconjugated | |
| Q60625 | |
| ICAM5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5 Leu31-Arg828 (Pro47Arg) Accession # Q60625 | |
| 100 μg | |
| Primary | |
| Detects mouse ICAM-5 in direct ELISAs. Detects mouse and rat ICAM-5 in Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ICAM-5 and recombinant mouse (rm) ICAM-2 is observed, 5% cross-reactivity with recombinant rat ICAM-1, rhICAM-1, and rhICAM-3 is observed and less than 1% cross-reactivity with rmICAM-1 and rmVCAM-1 is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7087 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto